Learn More
Medchemexpress LLC Nt1-o12b | 2739805-63-9 | 95.0% | 713.13 | 50 MG
SDP
Catalog No. 5000351318
Encompass_Preferred
NT1-O12B, an endogenous chemical and a neurotransmitter-derived lipidoid (NT-lipidoid), is an effective carrier for enhanced brain delivery of several blood-brain barrier (BBB)-impermeable cargos. Doping NT1-O12B into BBB-impermeable lipid nanoparticles (LNPs) gives the LNPs the ability to cross the BBB. NT-lipidoids formulation not only facilitate cargo crossing of the BBB, but also delivery of the cargo into neuronal cells for functional gene silencing or gene recombination.
  • Acts as an effective carrier for enhanced brain delivery of BBB-impermeable cargos
  • Doping into BBB-impermeable lipid nanoparticles (LNPs) enables them to cross the BBB
  • Facilitates cargo crossing of the BBB and delivery into neuronal cells
  • Supports functional gene silencing or gene recombination
Catalog No. 50-003-51318 Supplier Medchemexpress LLC Supplier No. HY13749950MG
NT1-O12B, an endogenous chemical and a neurotransmitter-derived lipidoid (NT-lipidoid), is an effective carrier for enhanced brain delivery of several blood-brain barrier (BBB)-impermeable cargos. Doping NT1-O12B into BBB-impermeable lipid nanoparticles (LNPs) gives the LNPs the ability to cross the BBB. NT-lipidoids formulation not only facilitate cargo crossing of the BBB, but also delivery of the cargo into neuronal cells for functional gene silencing or gene recombination.
  • Acts as an effective carrier for enhanced brain delivery of BBB-impermeable cargos
  • Doping into BBB-impermeable lipid nanoparticles (LNPs) enables them to cross the BBB
  • Facilitates cargo crossing of the BBB and delivery into neuronal cells
  • Supports functional gene silencing or gene recombination