Learn More
Hach Company Hach NT3100sc UV Nitrate Sensor, 2 mm path length (LXV448.99.21001)
Catalog No. NC3441558 Shop All Hach Company Products
Encompass
The Hach NT3100sc UV Nitrate Sensor (LXV448.99.21001) is a reagent-free, inline UV-absorbance sensor for continuous nitrate (NO3-N) monitoring in water and wastewater. It delivers low-level detection with sludge compensation, automatic wiping for reduced maintenance, and robust construction for process environments.
  • Reagent-free UV-absorbance measurement for nitrate.
  • 2 mm optical path length suitable for low-level detection.
  • Measurement range 0.05-50 mg/L NO₃-N.
  • Sludge-compensated, two-channel beam path for reliable readings.
  • Automatic wiper to minimise fouling and maintenance.
  • Stainless steel enclosure and quartz measuring window for durability.
  • Compact form factor: approx. 70 x 402 mm; weight approx. 4.8 kg with 10 m cable.
Catalog No. NC3441558 Supplier Hach Company Supplier No. LXV448.99.21001
Add to Cart
Add to Cart
Request a Quote
The Hach NT3100sc UV Nitrate Sensor (LXV448.99.21001) is a reagent-free, inline UV-absorbance sensor for continuous nitrate (NO3-N) monitoring in water and wastewater. It delivers low-level detection with sludge compensation, automatic wiping for reduced maintenance, and robust construction for process environments.
  • Reagent-free UV-absorbance measurement for nitrate.
  • 2 mm optical path length suitable for low-level detection.
  • Measurement range 0.05-50 mg/L NO₃-N.
  • Sludge-compensated, two-channel beam path for reliable readings.
  • Automatic wiper to minimise fouling and maintenance.
  • Stainless steel enclosure and quartz measuring window for durability.
  • Compact form factor: approx. 70 x 402 mm; weight approx. 4.8 kg with 10 m cable.