Learn More
Aviva Systems Biology Corp NT5C1A Antibody - C-terminal region (ARP60349_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60349_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NT5C1A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human NT5C1A. Alternate names: CN-I, CN-IA, CN1, CN1A, MGC119199, MGC119201ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-016