Learn More
Aviva Systems Biology Corp NT5C3 Antibody - middle region (ARP32185_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32185_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NT5C3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NT5C3. Alternate names: MGC27337, MGC87109, MGC87828, P5'N-1, PN-I, PSN1, UMPH, UMPH1, cN-III, NT5C3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54476