Learn More
Sino Biological NT5C3A / UMPH1 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 13904R009100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human NT5C3A / UMPH1 (rh NT5C3A / UMPH1; Catalog#13904-HNAE; NP_001002010; Met12-Leu297).
Catalog No. 50-224-7923