Learn More
Aviva Systems Biology Corp NT5DC1 Antibody - N-terminal region (ARP55511_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55511_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NT5DC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NT5DC1. Alternate names: C6orf200, LP2642, MGC131837, MGC24302, NT5C2L1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61528