Learn More
Aviva Systems Biology Corp NT5E Antibody - C-terminal region (ARP64972_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64972_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NT5E. It was validated on Western Blot. . Alternate names: CD73, E5NT, NT, NT5, NTE, eN, eNTApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-404