Learn More
Aviva Systems Biology Corp NT5M Antibody - N-terminal region (ARP57370_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57370_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NT5M. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human NT5M. Alternate names: dNT-2, dNT2, mdNApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-303