Learn More
Aviva Systems Biology Corp NT5M Antibody - middle region (ARP57371_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57371_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NT5M. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NT5M. Alternate names: dNT-2, dNT2, mdNApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54076