Learn More
Formulatrix NT8 LOW VOLUME TIP CS/3840
SDP
Catalog No. NC2605455
Encompass
Case of NT8 low volume tip. Contains 10 tip boxes x 384 tips per box = 3,840 tips
Catalog No. NC2605455 Supplier Formulatrix Supplier No. 363040_B
Case of NT8 low volume tip. Contains 10 tip boxes x 384 tips per box = 3,840 tips