Learn More
Cayman Chemical Antalarmin (hydrochloride), Purity: ≥98%, CAS: 220953-69-5, 50 mg
SDP
Catalog No. 502968706
Encompass_Preferred
Selective, nonpeptide antagonist of the CRH-R1 (Ki = 1 nM) that, consequently, reduces the release of corticotropin in response to chronic stress
Catalog No. 50-296-8706 Supplier Cayman Chemical Supplier No. 1514750 MG
Selective, nonpeptide antagonist of the CRH-R1 (Ki = 1 nM) that, consequently, reduces the release of corticotropin in response to chronic stress