Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci NTAQ1 Recombinant Protein
SDP

Catalog No. 16593795
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-593-795 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-593-795 Supplier ProSci Supplier No. 925060050
Only null left
Add to Cart
Add to Cart

Recombinant Protein

This recombinant protein can be used for biological assays. For research use only.

Human protein N-terminal glutamine amidohydrolase (WDYHV1) is an enzyme that in humans is encoded by the WDYHV1 gene, belongs to the NTAQ1 family.WDYHV1 mediates the side-chain deamidation of N-terminal glutamine residues to glutamate, which is an important step in N-end rule pathway of protein degradation. Conversion of the resulting N-terminal glutamine to glutamate renders the protein susceptible to arginylation, polyubiquitination and degradation as specified by the N-end rule. However,it does not act on substrates with internal or C-terminal glutamine andnon-glutamine residues in any position. With the exception of proline, all tested second-position residues on substrate peptides do not greatly influence the activity. In contrast, a proline at position 2, virtually abolishes deamidation of N-terminal glutamine.

Specifications

Accession Number Q96HA8
Gene ID (Entrez) 55093
Molecular Weight (g/mol) 49.8 kDa
Quantity 0.05 mg
Source E. coli
Common Name WDYHV motif containing 1
Gene Symbol WDYHV1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag N-GST tag
Sequence Met1-Cys205
Content And Storage Store at -20°C, stable for 6 months after receipt. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Purity or Quality Grade >95% as determined by reducing SDS-PAGE. Endotoxin level less than 0.1 ng/μg (1 IEU/μg) as determined by LAL test.
Synonym Protein N-terminal glutamine amidohydrolase, WDYHV1, Protein NH2-terminal glutamine deamidase, N-terminal Gln amidase, Nt(Q)-amidase, C8orf32, NTAQ1, REF: C1009
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.