Learn More
CPC Scientific H-DPhe-Cys-Tyr-DTrp-Orn-Thr-Pen-Thr-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific SOMA005B
Encompass
[Cys2, Tyr3,Orn5,Pen7]. This somatostatin analog is a very potent and highly selective ligand for m-opioid receptors.ONE-LETTER SEQUENCE: DPhe-CY-DTrp-Orn-T-Pen-T-NH2 (Cys2 and Pen7 Bridge)MOLECULAR FORMULA: C50H68N11O11S2MOLECULAR WEIGHT:1063.3STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [103429-31-8], RESEARCH AREA: PainREFERENCES: K.N.Hawkins et al., J. Pharmacol. Exp. Ther., 248, 73 (1989); K.Gulya et al., Eur. J. Pharmacol., 150, 355 (1988); L.F. Tseng et al., The Journal of Pharmacol. and Experim. Therap., 280, 600 (1997)
Catalog No. 50-211-0428