Learn More
ACROBiosystems Human NTB-A Protein, His Tag, Lyophilized, 1 mg
Catalog No. 501657824
Encompass_Preferred
Human NTB-A, also known as SLAMF6, is a member of the SLAM family of receptors. Expressed from human HEK293 cells, this protein includes amino acids Gln 22 to Met 226 and features a polyhistidine tag at the C-terminus. It is supplied as a lyophilized powder and has been verified by SEC-MALS for quality.
  • Greater than 95% purity as determined by SDS-PAGE and SEC-MALS
  • Calculated molecular weight of 25.0 kDa, migrating as 35-45 kDa under reducing conditions due to glycosylation
  • Endotoxin level less than 1.0 EU per μg
  • Formulated in PBS, pH 7.4 with trehalose as a protectant
  • Stable for 12 months at -20°C to -70°C in lyophilized state
  • Shipped at ambient temperature
Catalog No. 50-165-7824 Supplier ACROBiosystems Supplier No. NTAH52HA1MG
Human NTB-A, also known as SLAMF6, is a member of the SLAM family of receptors. Expressed from human HEK293 cells, this protein includes amino acids Gln 22 to Met 226 and features a polyhistidine tag at the C-terminus. It is supplied as a lyophilized powder and has been verified by SEC-MALS for quality.
  • Greater than 95% purity as determined by SDS-PAGE and SEC-MALS
  • Calculated molecular weight of 25.0 kDa, migrating as 35-45 kDa under reducing conditions due to glycosylation
  • Endotoxin level less than 1.0 EU per μg
  • Formulated in PBS, pH 7.4 with trehalose as a protectant
  • Stable for 12 months at -20°C to -70°C in lyophilized state
  • Shipped at ambient temperature