Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED ON-TARGETplus Non-targeting Control siRNAs #2, 5nmol
Catalog No. NC1622123
Encompass
ON-TARGETplus Non-targeting Control siRNAs are the ideal negative control for RNAi experiments in human, mouse, and rat cells. ON-TARGETplus Non-targeting siRNAs were designed to leverage seed-region optimization and patented ON-TARGETplus modification patterns to have fewer off-targets than traditionally designed, unmodified negative control siRNAs. Changes in mRNA or protein levels in cells treated with these controls reflect a baseline cellular response that can be compared to the levels in cells treated with target-specific siRNA.
Catalog No. NC1622123 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. D0018100205
ON-TARGETplus Non-targeting Control siRNAs are the ideal negative control for RNAi experiments in human, mouse, and rat cells. ON-TARGETplus Non-targeting siRNAs were designed to leverage seed-region optimization and patented ON-TARGETplus modification patterns to have fewer off-targets than traditionally designed, unmodified negative control siRNAs. Changes in mRNA or protein levels in cells treated with these controls reflect a baseline cellular response that can be compared to the levels in cells treated with target-specific siRNA.