Learn More
ATCC NTERA-2 cl.D1; Embryonal Carcinoma; Human (Homo sapiens)
Catalog No. 502384592
Encompass
NTERA-2 cl.D1 [NT2/D1] is a cell line exhibiting epithelial-like, differentiation changes phenotype, morphology that was isolated in 1980 from the testis of a White, 22-year-old, male patient with carcinoma, malignant pluripotent embryonal. This cell line was deposited by PW Andrews and can be used cancer research.
Catalog No. 50-238-4592 Supplier ATCC Supplier No. CRL1973
NTERA-2 cl.D1 [NT2/D1] is a cell line exhibiting epithelial-like, differentiation changes phenotype, morphology that was isolated in 1980 from the testis of a White, 22-year-old, male patient with carcinoma, malignant pluripotent embryonal. This cell line was deposited by PW Andrews and can be used cancer research.