Learn More
Cayman Chemical Anthraflavic Acid, Purity: ≥95%, CAS: 84-60-6, 10 g
SDP
Catalog No. 502918385
Encompass_Preferred
An anthraquinone with diverse biological activities; inhibits α-amylase in a cell-free assay (IC50 = 198.3 nM) and cytochrome P450 activity induced by a mixture of PCBs in rat hepatic microsomes; binds to ERα and ERβ (Kis = 0.31 and 0.69 µM, respectively); cytotoxic against a panel of six cancer cell lines (IC50s = 156-241 µg/ml); inhibits Glu-P-1-induced mutagenicity in the Ames test using S. typhimurium at 50 µM; topical application reduces the number of tumors per mouse and the number of mice with tumors in a model of two-stage carcinogenesis initiated by DMBA and promoted by TPA; has been used as an anolyte in redox flow batteries
Catalog No. 50-291-8385 Supplier Cayman Chemical Supplier No. 3924610 G
An anthraquinone with diverse biological activities; inhibits α-amylase in a cell-free assay (IC50 = 198.3 nM) and cytochrome P450 activity induced by a mixture of PCBs in rat hepatic microsomes; binds to ERα and ERβ (Kis = 0.31 and 0.69 µM, respectively); cytotoxic against a panel of six cancer cell lines (IC50s = 156-241 µg/ml); inhibits Glu-P-1-induced mutagenicity in the Ames test using S. typhimurium at 50 µM; topical application reduces the number of tumors per mouse and the number of mice with tumors in a model of two-stage carcinogenesis initiated by DMBA and promoted by TPA; has been used as an anolyte in redox flow batteries