Learn More
Cayman Chemical ntI-CItrulIn Monclonal n 100ug
SDP
Catalog No. 502988113
Encompass_Preferred
Immunogen: Citrulline-containing peptide conjugated to keyhole limpet hemocyanin • Host: Mouse • Species Reactivity: (+) Species independent • Cross Reactivity (+) Citrullinated proteins; (-) Native proteins • Applications: ELISA, IHC, IP, and WB
Catalog No. 50-298-8113 Supplier Cayman Chemical Supplier No. 30773100 UG
Immunogen: Citrulline-containing peptide conjugated to keyhole limpet hemocyanin • Host: Mouse • Species Reactivity: (+) Species independent • Cross Reactivity (+) Citrullinated proteins; (-) Native proteins • Applications: ELISA, IHC, IP, and WB