Learn More
Cayman Chemical ntI-EP2 Receptr SureLIght50ug
SDP
Catalog No. 502965620
Encompass_Preferred
An N-hydroxysuccinimide ester of the nitroxide spin label tempyo an active acylating agent that preferentially targets a-amino groups used to spin label proteins and aminoacylated RNA species
Catalog No. 50-296-5620 Supplier Cayman Chemical Supplier No. 1668450 UG
An N-hydroxysuccinimide ester of the nitroxide spin label tempyo an active acylating agent that preferentially targets a-amino groups used to spin label proteins and aminoacylated RNA species