Learn More
Cayman Chemical ntI-H2AXphospho-Ser139 R 100ug
SDP
Catalog No. 502980766
Encompass_Preferred
Immunogen: A phospho-peptide corresponding to phospho-histone • Host: Rabbit • Species Reactivity: (+) All species • Applications: ELISA, ICC, Multiplex, and WB • A probe for immunochemical detection of phospho-γH2AXSer139
Catalog No. 50-298-0766 Supplier Cayman Chemical Supplier No. 20721100 UG
Immunogen: A phospho-peptide corresponding to phospho-histone • Host: Rabbit • Species Reactivity: (+) All species • Applications: ELISA, ICC, Multiplex, and WB • A probe for immunochemical detection of phospho-γH2AXSer139