Learn More
Cayman Chemical ntI-Inflamatry Screeng L 25uL
SDP
Catalog No. 502991084
Encompass_Preferred
An internal standard for the quantification of decanoic acid by GC- or LC-MS
Catalog No. 50-299-1084 Supplier Cayman Chemical Supplier No. 3153025 UL
An internal standard for the quantification of decanoic acid by GC- or LC-MS