Learn More
Cayman Chemical ntI-InterleukIn-1ahumn E 1ea
SDP
Catalog No. 502893835
Encompass_Preferred
An unsaturated ionizable cationic lipid has been used in the generation of LNPs for the delivery of mRNA in vivo intravenous administration of LNPs containing lipid 29d and encapsulating mRNA encoding a luciferase reporter induce bioluminescence in the lungs and spleen of mice
Catalog No. 50-289-3835 Supplier Cayman Chemical Supplier No. 4833021 EA
An unsaturated ionizable cationic lipid has been used in the generation of LNPs for the delivery of mRNA in vivo intravenous administration of LNPs containing lipid 29d and encapsulating mRNA encoding a luciferase reporter induce bioluminescence in the lungs and spleen of mice