Learn More
Cayman Chemical ntI-InterleukIn-1ahumn E 5ea
SDP
Catalog No. 502893870
Encompass_Preferred
An unsaturated ionizable cationic lipid has been used in the generation of LNPs for the delivery of mRNA in vivo intravenous administration of LNPs containing lipid 29d and encapsulating mRNA encoding a luciferase reporter induce bioluminescence in the lungs and spleen of mice
Catalog No. 50-289-3870 Supplier Cayman Chemical Supplier No. 4833025 EA
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
An unsaturated ionizable cationic lipid has been used in the generation of LNPs for the delivery of mRNA in vivo intravenous administration of LNPs containing lipid 29d and encapsulating mRNA encoding a luciferase reporter induce bioluminescence in the lungs and spleen of mice