Learn More
Sigma Aldrich Fine Chemicals Biosciences Anti-LC3B Polyclonal Antibody, from Rabbit | CAS No: N/A | MDL No: N/A | 1mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L7543100UL
Encompass
Anti-LC3B polyclonal antibody | Purity: N/A | Mol Wt: N/A | CAS No: N/A | MDL No: N/A | 1mLThis rabbit polyclonal antibody recognizes the 14.7 kDa LC3B protein, an autophagosomal membrane marker, enabling immunoblotting detection across human, mouse and rat species. The MAP1LC3B antibody is supplied in PBS with sodium azide preservative at ~1 mg/mL concentration for monitoring autophagy.
Catalog No. NC2599296