Learn More
Cayman Chemical ntI5methyl CytosIn RabbIt 50ug
SDP
Catalog No. 502982656
Encompass_Preferred
Immunogen: BSA-conjugated 5-mC • Host: Rabbit • Species Reactivity: Species-independent • Cross Reactivity: (+) 5-mC in ssDNA and dsDNA; (-) Non-methylated cytosine, 5-hmC • Applications: ELISA, dot blot, FC, ICC, MeDIP
Catalog No. 50-298-2656 Supplier Cayman Chemical Supplier No. 2072250 UG
Immunogen: BSA-conjugated 5-mC • Host: Rabbit • Species Reactivity: Species-independent • Cross Reactivity: (+) 5-mC in ssDNA and dsDNA; (-) Non-methylated cytosine, 5-hmC • Applications: ELISA, dot blot, FC, ICC, MeDIP