Learn More
Cayman Chemical ntIBeta 2MCroglobuln HRP 1.5mL
SDP
Catalog No. 502894610
Encompass_Preferred
Anti-Beta 2-Microglobulin HRP Conjugate has been tested and formulated to work exclusively with Cayman’s MHC Class I (human) ELISA Kit (Item No.502060).
Catalog No. 50-289-4610 Supplier Cayman Chemical Supplier No. 5020621.5 ML
Anti-Beta 2-Microglobulin HRP Conjugate has been tested and formulated to work exclusively with Cayman’s MHC Class I (human) ELISA Kit (Item No.502060).