Learn More
Cayman Chemical Antide (acetate), Purity: ≥98%, CAS: 625092-10-6, 10 mg
SDP
Catalog No. 502991911
Encompass_Preferred
A GnRHR antagonist; inhibits GnRH-induced secretion of LH, FSH, and gonadotropin in isolated rat pituitary cells (0.001-1,000 nM) but does not induce histamine release in isolated rat mast cells (EC50 = >300 µg/ml); reduces serum LH and FSH levels in orchidectomized cynomolgus monkeys at 250 µg/kg; inhibits ovulation in rats at 2 µg/animal; reduces the intensity and duration of haloperidol-induced catalepsy at 10 µg/animal
Catalog No. 50-299-1911 Supplier Cayman Chemical Supplier No. 3148610 MG
A GnRHR antagonist; inhibits GnRH-induced secretion of LH, FSH, and gonadotropin in isolated rat pituitary cells (0.001-1,000 nM) but does not induce histamine release in isolated rat mast cells (EC50 = >300 µg/ml); reduces serum LH and FSH levels in orchidectomized cynomolgus monkeys at 250 µg/kg; inhibits ovulation in rats at 2 µg/animal; reduces the intensity and duration of haloperidol-induced catalepsy at 10 µg/animal