Learn More
Cayman Chemical ntIDNP PolycLNal ANTIBDY 500uL
SDP
Catalog No. 502936783
Encompass_Preferred
An antispasmodic agent an inhibitor of AChE (Kis 36 and 45 M for bovine erythrocyte and electric eel enzyme respectively) inhibits acetylcholine-induced contractions of isolated guinea pig ileum at 2.5 M
Catalog No. 50-293-6783 Supplier Cayman Chemical Supplier No. 25864500 UL
An antispasmodic agent an inhibitor of AChE (Kis 36 and 45 M for bovine erythrocyte and electric eel enzyme respectively) inhibits acetylcholine-induced contractions of isolated guinea pig ileum at 2.5 M