Learn More
Cayman Chemical ntIlImIcInsulfate 10mg
SDP
Catalog No. 502936956
Encompass_Preferred
An aminoglycoside antibiotic; active against S. aureus, N. gonorrhoeae, E. coli, P. mirabilis, and P. aeruginosa (MICs = 0.4, 3.1, 0.8, 1.6, and 0.4 UG/ml, respectively) as well as Enterococci, Enterobacter, Citrobacter, and Klebsiella species (MICs = 3.1, 0.4, 0.8, and ≤0.2 UG/ml, respectively); induces nephrotoxicity in rats at 50-150 mg/kg; induces neuromuscular block in isolated diaphragm and soleus muscles in rabbits (ED50s = 18.5 and 62.2 mg/kg, respectively)
Catalog No. 50-293-6956 Supplier Cayman Chemical Supplier No. 2666610 MG
An aminoglycoside antibiotic; active against S. aureus, N. gonorrhoeae, E. coli, P. mirabilis, and P. aeruginosa (MICs = 0.4, 3.1, 0.8, 1.6, and 0.4 UG/ml, respectively) as well as Enterococci, Enterobacter, Citrobacter, and Klebsiella species (MICs = 3.1, 0.4, 0.8, and ≤0.2 UG/ml, respectively); induces nephrotoxicity in rats at 50-150 mg/kg; induces neuromuscular block in isolated diaphragm and soleus muscles in rabbits (ED50s = 18.5 and 62.2 mg/kg, respectively)