Learn More
Cayman Chemical ntImycIn A Complex 100mg
SDP
Catalog No. 502913119
Encompass_Preferred
A bacterial metabolite complex; induces the synthesis of carotenoids in M. marinum at 3.3-160 µM; has teleocidal activity against goldfish at 25-125 ppb
Catalog No. 50-291-3119 Supplier Cayman Chemical Supplier No. 34799100 MG
A bacterial metabolite complex; induces the synthesis of carotenoids in M. marinum at 3.3-160 µM; has teleocidal activity against goldfish at 25-125 ppb