Learn More
Cayman Chemical Antimycin A Complex, Purity: ≥95% (mixture of subunits), CAS: 1397-94-0, 50 mg
SDP
Catalog No. 502911176
Encompass_Preferred
A bacterial metabolite complex; induces the synthesis of carotenoids in M. marinum at 3.3-160 µM; has teleocidal activity against goldfish at 25-125 ppb
Catalog No. 50-291-1176 Supplier Cayman Chemical Supplier No. 3479950 MG
A bacterial metabolite complex; induces the synthesis of carotenoids in M. marinum at 3.3-160 µM; has teleocidal activity against goldfish at 25-125 ppb