Learn More
Cayman Chemical ntImycIn A1 2.5mg
SDP
Catalog No. 502983448
Encompass_Preferred
An active component of the antimycin A complex; inhibits electron transport; induces the synthesis of carotenoids in M. marinum at 3.3-160 µM; inhibits ATP-citrate lyase (Ki = 29.5 µM)
Catalog No. 50-298-3448 Supplier Cayman Chemical Supplier No. 194332.5 MG
An active component of the antimycin A complex; inhibits electron transport; induces the synthesis of carotenoids in M. marinum at 3.3-160 µM; inhibits ATP-citrate lyase (Ki = 29.5 µM)