Learn More
Cayman Chemical Antimycin A2, Purity: ≥85%, CAS: 27220-57-1, 2.5 mg
SDP
Catalog No. 502987683
Encompass_Preferred
A component of the antimycin A complex and an inhibitor of ATP-citrate lyase (Ki = 4.2 µM)
Catalog No. 50-298-7683 Supplier Cayman Chemical Supplier No. 219972.5 MG
 Restricted Item
Add to Cart
Add to Cart
A component of the antimycin A complex and an inhibitor of ATP-citrate lyase (Ki = 4.2 µM)