Learn More
Cayman Chemical ntImycIn A4 2.5mg
SDP
Catalog No. 502987890
Encompass_Preferred
A component of the antimycin A complex and an inhibitor of ATP-citrate lyase (Ki = 64.8 µM)
Catalog No. 50-298-7890 Supplier Cayman Chemical Supplier No. 219992.5 MG
Add to Cart
Add to Cart
A component of the antimycin A complex and an inhibitor of ATP-citrate lyase (Ki = 64.8 µM)