Learn More
Cayman Chemical ntIOvalbumIn IgEmouse EL 200ng
SDP
Catalog No. 502942505
Encompass_Preferred
An internal standard for the quantification of AEA by GC- or LC-MS.
Catalog No. 50-294-2505 Supplier Cayman Chemical Supplier No. 400844200 NG
An internal standard for the quantification of AEA by GC- or LC-MS.