Learn More
Cayman Chemical ntISARSCoV2 SpIke Glycopr 1ea
SDP
Catalog No. 502896949
Encompass_Preferred
Anti-SARS-CoV-2 Spike Glycoprotein RBD IgG Negative Control has been tested and formulated to work exclusively with Cayman’s Anti-SARS-CoV-2 Spike Glycoprotein RBD IgG ELISA Kit (Item No. 502111).
Catalog No. 50-289-6949 Supplier Cayman Chemical Supplier No. 5021151 EA
Anti-SARS-CoV-2 Spike Glycoprotein RBD IgG Negative Control has been tested and formulated to work exclusively with Cayman’s Anti-SARS-CoV-2 Spike Glycoprotein RBD IgG ELISA Kit (Item No. 502111).