Learn More
Cayman Chemical SARS-CoV-2 Spike Glycoprotein RBD Reagent, 50 dtn
SDP
Catalog No. 502896041
Encompass_Preferred
Anti-SARS-CoV-2 Spike Glycoprotein RBD Reagent has been tested and formulated to work exclusively with Cayman’s Anti-SARS-CoV-2 Spike Glycoprotein RBD IgG ELISA Kit (Item No. 502111).
Catalog No. 50-289-6041 Supplier Cayman Chemical Supplier No. 50211150 DTN
Anti-SARS-CoV-2 Spike Glycoprotein RBD Reagent has been tested and formulated to work exclusively with Cayman’s Anti-SARS-CoV-2 Spike Glycoprotein RBD IgG ELISA Kit (Item No. 502111).