Learn More
Cayman Chemical ntIvIral Screeng LIbrary 50uL
SDP
Catalog No. 502992019
Encompass_Preferred
An inhibitor of Tie2 and VEGFR2 (IC50s 1 and 3 nM respectively in a HTRF assay) inhibits angiopoietin 1-induced Tie2 autophosphorylation in EA hy926 human endothelial cells (IC50 10 nM)
Catalog No. 50-299-2019 Supplier Cayman Chemical Supplier No. 3039050 UL
An inhibitor of Tie2 and VEGFR2 (IC50s 1 and 3 nM respectively in a HTRF assay) inhibits angiopoietin 1-induced Tie2 autophosphorylation in EA hy926 human endothelial cells (IC50 10 nM)