Learn More
Aviva Systems Biology Corp NTN4 Antibody - N-terminal region (ARP49645_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49645_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NTN4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NTN4. Alternate names: FLJ23180, PRO3091ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53594