Learn More
Aviva Systems Biology Corp NTNG2 Antibody - middle region (ARP60352_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60352_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NTNG2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human NTNG2. Alternate names: KIAA0625, KIAA1857, LHLL9381, Lmnt2, MGC21884, NTNG1, bA479K20.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-017