Learn More
Cayman Chemical nTosIs asy KIt 1ea
SDP
Catalog No. 502897047
Encompass_Preferred
Cayman’s NETosis Assay Kit provides a simple and fast method for studying the process of NETosis ex vivo. Notably, Cayman’s NETosis Assay Kit does not depend upon the DNA component of NETs, as DNA release can occur independently of NETosis. In this kit, primary neutrophils are stimulated to release NETs with either PMA or a calcium ionophore (both included). As shown in Figure 1, unbound neutrophil elastase is washed away following NET generation. Following digestion of NET DNA by S7 nuclease, the supernatant containing neutrophil elastase is added to a substrate, which is selectively cleaved by neutrophil elastase to yield a 4-nitroaniline product that absorbs light at 405 nm. A sufficient amount of EAch reagent is provided to test and analyze up to 80 individual samples or 24 samples in duplicate.
Catalog No. 50-289-7047 Supplier Cayman Chemical Supplier No. 6010101 EA
Add to Cart
Add to Cart
Cayman’s NETosis Assay Kit provides a simple and fast method for studying the process of NETosis ex vivo. Notably, Cayman’s NETosis Assay Kit does not depend upon the DNA component of NETs, as DNA release can occur independently of NETosis. In this kit, primary neutrophils are stimulated to release NETs with either PMA or a calcium ionophore (both included). As shown in Figure 1, unbound neutrophil elastase is washed away following NET generation. Following digestion of NET DNA by S7 nuclease, the supernatant containing neutrophil elastase is added to a substrate, which is selectively cleaved by neutrophil elastase to yield a 4-nitroaniline product that absorbs light at 405 nm. A sufficient amount of EAch reagent is provided to test and analyze up to 80 individual samples or 24 samples in duplicate.