Learn More
Cayman Chemical nTosIs Imagng asy KIt 96Wel
SDP
Catalog No. 502896992
Encompass_Preferred
Cayman’s NETosis Imaging Kit provides a simple staining protocol for visualizing the process of NETosis kinetically ex vivo and in vitro. A cell-permeable DNA dye allows visualization of the dynamic changes of the nucleus during the process of NETosis, while a cell-impermeable dye detects the extruded DNA. Combining the fluorescence visualization of these dyes with brightfield imaging in a high-content platform allows for exquisite differentiation between different modes of cell death. The amount of EAch reagent provided is sufficient for two 96-well plates.
Catalog No. 50-289-6992 Supplier Cayman Chemical Supplier No. 60175096 WELL
Cayman’s NETosis Imaging Kit provides a simple staining protocol for visualizing the process of NETosis kinetically ex vivo and in vitro. A cell-permeable DNA dye allows visualization of the dynamic changes of the nucleus during the process of NETosis, while a cell-impermeable dye detects the extruded DNA. Combining the fluorescence visualization of these dyes with brightfield imaging in a high-content platform allows for exquisite differentiation between different modes of cell death. The amount of EAch reagent provided is sufficient for two 96-well plates.