Learn More
Apexbio Technology LLC NTP set, 100 mM solution,1 Set (4x250uL)
SDP
Catalog No. 502240947
Encompass_Preferred
This product is a set of 4 solutions of ATP, UTP, GTP and CTP (in 4 tubes) and can be used for a variety of related applications in molecular biology, such as in vitro transcription, RNA amplification, siRNA synthesis, etc. It can also be used as a reaction substrate or coenzyme for a variety of enzymes. This product is a colourless and transparent aqueous solution prepared from trisodium salts of ATP, UTP, GTP and CTP with a purity of greater than or equal to 99%. It has a pH (25 °C) value of 7.0 ± 0.1, a concentration value of 100 mM and is free from DNase and RNase contamination.Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0947 Supplier Apexbio Technology LLC Supplier No. K10491
This product is a set of 4 solutions of ATP, UTP, GTP and CTP (in 4 tubes) and can be used for a variety of related applications in molecular biology, such as in vitro transcription, RNA amplification, siRNA synthesis, etc. It can also be used as a reaction substrate or coenzyme for a variety of enzymes. This product is a colourless and transparent aqueous solution prepared from trisodium salts of ATP, UTP, GTP and CTP with a purity of greater than or equal to 99%. It has a pH (25 °C) value of 7.0 ± 0.1, a concentration value of 100 mM and is free from DNase and RNase contamination.Other sizes are also available. Please inqury us for quote.