Learn More
TARGETMOL CHEMICALS INC NTP42 5MG
SDP
Catalog No. 502581473
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NTP42 is an antagonist of thromboxane A2 (TXA2) receptor(IC50 of 3.278 nM)purity: 99%
Catalog No. 50-258-1473 Supplier TARGETMOL CHEMICALS INC Supplier No. T122685MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NTP42 is an antagonist of thromboxane A2 (TXA2) receptor(IC50 of 3.278 nM)purity: 99%