Learn More
Medchemexpress LLC HY-129851 5mg Medchemexpress, NTP42 CAS:2055599-51-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1298515MG
Encompass_Preferred
Medchemexpress, HY-129851 5mg NTP42 CAS:2055599-51-2 NTP42 is a thromboxane A2 (TXA2) receptor antagonist with an IC50 of 3.278 nM for antagonizing T prostanoid receptor (TP)- mediated [Ca2+] mobilization following stimulation of cells with the alternative TP agonist U46609. NTP42 can be used for the treatment of pulmonary arterial hypertension (PAH). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9819