Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ntpa Tfa | 99.8% | 1773.36 | 1 MG
SDP

Catalog No. 5000396349
Encompass_Preferred

NTPA TFA is a polycarboxylic acid ligand that participates in the coordination reaction with metal ions to form complexes.

  • High purity: 99.80%
  • Molecular weight: 1773.36 (free base)
  • Appearance: solid
  • Color: white to off-white
  • Sequence: C17H35CONH-Pro-Lys-Lys-Lys-Arg-Lys-Val-Arg-Arg-Arg-Arg-NH2
  • Soluble in DMSO (50 mg/mL)

Catalog No. 50-003-96349 Supplier Medchemexpress LLC Supplier No. HYP64431MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NTPA TFA is a polycarboxylic acid ligand that participates in the coordination reaction with metal ions to form complexes.

  • High purity: 99.80%
  • Molecular weight: 1773.36 (free base)
  • Appearance: solid
  • Color: white to off-white
  • Sequence: C17H35CONH-Pro-Lys-Lys-Lys-Arg-Lys-Val-Arg-Arg-Arg-Arg-NH2
  • Soluble in DMSO (50 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.