Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NTPA TFA | 99.8% | 1773.36 (free base) | 5 MG
SDP

Catalog No. 5000398046
Encompass_Preferred

NTPA TFA is a polycarboxylic acid ligand that participates in the coordination reaction with metal ions to form complexes. It is intended for research use only.

  • Purity of 99.80%
  • Molecular weight of 1773.36 (free base)
  • Formula: C82H161N31O12.xC2HF3O2
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Sealed storage, away from moisture. Powder: -80°C for 2 years, -20°C for 1 year. In solvent: -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture)
  • Solubility: In vitro: DMSO : 50 mg/mL (need ultrasonic; hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)

Catalog No. 50-003-98046 Supplier Medchemexpress LLC Supplier No. HYP64435MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NTPA TFA is a polycarboxylic acid ligand that participates in the coordination reaction with metal ions to form complexes. It is intended for research use only.

  • Purity of 99.80%
  • Molecular weight of 1773.36 (free base)
  • Formula: C82H161N31O12.xC2HF3O2
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Sealed storage, away from moisture. Powder: -80°C for 2 years, -20°C for 1 year. In solvent: -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture)
  • Solubility: In vitro: DMSO : 50 mg/mL (need ultrasonic; hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.