Learn More
ABclonal Technology NTPCR Rabbit pAb
SDP
Catalog No. 5000108631
Encompass_Preferred
The protein encoded by this gene is a non-specific nucleoside triphosphatase that is slow-acting in vitro. This gene is overexpressed in many tumor tissues, and while it is not essential for the cell, overexpression is cytotoxic. However, the cytotoxicity is not related to its triphosphatase activity.
Catalog No. 50-001-08631 Supplier ABclonal Technology Supplier No. A14944500UL
The protein encoded by this gene is a non-specific nucleoside triphosphatase that is slow-acting in vitro. This gene is overexpressed in many tumor tissues, and while it is not essential for the cell, overexpression is cytotoxic. However, the cytotoxicity is not related to its triphosphatase activity.