Learn More
Medchemexpress LLC NTRC 0066-0 | 1817791-73-3 | 99.0% | 565.71 g/mol | C33H39N7O2 | 1 ML
SDP
Catalog No. 5000215340
Encompass_Preferred
NTRC 0066-0 is a potent, selective threonine tyrosine kinase (TTK/MPS1) inhibitor (IC50 = 0.9 nM) supplied for preclinical research. The compound is available as a 10 mM solution in DMSO (1 mL) and as solid quantities, and is characterized by a molecular weight of 565.71 g/mol, formula C33H39N7O2, and CAS 1817791-73-3.
  • Sub-nanomolar potency against TTK (IC50 = 0.9 nM).
  • High reported purity (>99%).
  • Supplied as a 10 mM solution in DMSO for convenient dosing.
  • Documented solubility and storage conditions for reliable handling.
  • Suitable for cancer biology and kinase inhibitor research applications.
Catalog No. 50-002-15340 Supplier Medchemexpress LLC Supplier No. HY10002410MM 1ML
NTRC 0066-0 is a potent, selective threonine tyrosine kinase (TTK/MPS1) inhibitor (IC50 = 0.9 nM) supplied for preclinical research. The compound is available as a 10 mM solution in DMSO (1 mL) and as solid quantities, and is characterized by a molecular weight of 565.71 g/mol, formula C33H39N7O2, and CAS 1817791-73-3.
  • Sub-nanomolar potency against TTK (IC50 = 0.9 nM).
  • High reported purity (>99%).
  • Supplied as a 10 mM solution in DMSO for convenient dosing.
  • Documented solubility and storage conditions for reliable handling.
  • Suitable for cancer biology and kinase inhibitor research applications.