Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NTRC-844 | 1811503-67-9 | 95.6% | 489.54 g·mol⁻1 | C28H28FN3O4 | 100MG
SDP

Catalog No. 5000237791
Encompass_Preferred

NTRC-844 is a small-molecule, selective antagonist of neurotensin receptor type 2 (NTS2) used in preclinical analgesia research. The compound shows an EC50 of 238 nM at NTS2 and is supplied as a solid for laboratory research.

  • Selective NTS2 antagonist with reported EC50 of 238 nM.
  • Suitable for preclinical analgesic and neurotensin receptor research.
  • Molecular formula C28H28FN3O4; molecular weight 489.54 g·mol⁻1.
  • Purity reported by HPLC as 95.6%.
  • Supplied in small milligram quantities for laboratory use.
  • Characterization confirmed by LC-MS.

Catalog No. 50-002-37791 Supplier Medchemexpress LLC Supplier No. HY124831100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NTRC-844 is a small-molecule, selective antagonist of neurotensin receptor type 2 (NTS2) used in preclinical analgesia research. The compound shows an EC50 of 238 nM at NTS2 and is supplied as a solid for laboratory research.

  • Selective NTS2 antagonist with reported EC50 of 238 nM.
  • Suitable for preclinical analgesic and neurotensin receptor research.
  • Molecular formula C28H28FN3O4; molecular weight 489.54 g·mol⁻1.
  • Purity reported by HPLC as 95.6%.
  • Supplied in small milligram quantities for laboratory use.
  • Characterization confirmed by LC-MS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.